Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min

Chemical Identifiers

Chemical Name or Material Rhodium(III) iodide
CAS 15492-38-3
Formula Weight 483.62
MDL Number MFCD00016311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1102901

  SDS
alfa aesar™
1102901
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1102906

  SDS
alfa aesar™
1102906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1102903

  SDS
alfa aesar™
1102903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 483.62
Color Black
CAS 15492-38-3
Chemical Name or Material Rhodium(III) iodide
Assay Percent Notes (metals basis)
Concentration Rh 20.8% min.
Odor Odorless
Quantity 0.25g
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00016311
Health Hazard 1 Warning
RTECS Number VI9315000
TSCA TSCA
Physical Form Crystalline
Assay 99.9%
Sensitivity Hygroscopic
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.