missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min

Manufacturer:  Alfa Aesar™ 1102903

 View more versions of this product

Catalog No. AA1102903



Specifications

Specifications

Rhodium(III) iodide
(metals basis)
483.62
99.9%
Odorless
Black
15492-38-3
MFCD00016311
Crystalline
Rh 20.8% min.
Hygroscopic
1g

Chemical Identifiers

15492-38-3
MFCD00016311
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VI9315000

TSCA : TSCA

Documents