missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) chloride solution, 99.5% (metals basis), Rh 10% w/w (cont. Rh)

Manufacturer:  Alfa Aesar™ 46523NJ

 View more versions of this product

Catalog No. AA46523NJ



Specifications

Specifications

Rhodium (III) chloride solution
(metals basis)
RhCl3
MFCD00011204
14,8188
Liquid
Brown-Red
10049-07-7
Cl3Rh
Rh 10% w/w (cont. Rh)
UN3264
209.26
99.5%
0.5g

Chemical Identifiers

10049-07-7
Cl3Rh
MFCD00011204
Safety and Handling

Safety and Handling

GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : VI9275000

TSCA : TSCA

SDS Documents