missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium(III) chloride hydrate, Rh 38.5-45.5%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1103201

 View more versions of this product

Catalog No. AA1103201



Specifications

Specifications

Rhodium(III) chloride hydrate
20765-98-4
RhCl3·xH2O
Dark red crystals
209.26 (Anhydrous)
Red
Rh 38.5-45.5%
38.5 to 45.5%
MFCD00149839
Rhodium trichloride hydrate
Crystalline
0.25g

Chemical Identifiers

20765-98-4
RhCl3·xH2O
MFCD00149839
Rhodium trichloride hydrate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.

H318-H290-H341-H302: Causes serious eye damage. May be corrosive to metals. Suspected of causing genetic defects. Harmful if swallowed.

Documents