Please login to your online account to display your discounted pricing

Rhodium(III) chloride hydrate, Rh 38.5-45.5%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Rhodium(III) chloride hydrate
Name Note Rh 38.5-45.5%
CAS 20765-98-4
Physical Form Crystalline
Assay Percent Range 38.5 to 45.5%
Molecular Formula RhCl3·xH2O
Formula Weight 209.26 (anhydrous)
MDL Number MFCD00149839
Synonym Rhodium trichloride hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1103201

  SDS
alfa aesar
1103201
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103203

  SDS
alfa aesar
1103203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1103206

  SDS
alfa aesar
1103206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Rhodium(III) chloride hydrate
Name Note Rh 38.5-45.5%
CAS 20765-98-4
Health Hazard 1 H318-H290-H341-H302: Causes serious eye damage. May be corrosive to metals. Suspected of causing genetic defects. Harmful if swallowed.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.
Physical Form Crystalline
Assay Percent Range 38.5 to 45.5%
Molecular Formula RhCl3·xH2O
Appearance Dark red crystals
Formula Weight 209.26 (Anhydrous)
MDL Number MFCD00149839
Synonym Rhodium trichloride hydrate
Color Dark red
Quantity 0.25g
Safety and Handling

H318-H290-H341-H302: Causes serious eye damage. May be corrosive to metals. Suspected of causing genetic defects. Harmful if swallowed.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.