Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min

Chemical Identifiers

Chemical Name or Material Rhodium(III) chloride
CAS 10049-07-7
Name Note Anhydrous
Linear Formula RhCl3
Formula Weight 209.26
MDL Number MFCD00011204
Merck Index 14,8188
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1181501

 
alfa aesar™
1181501
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1181503

 
alfa aesar™
1181503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1181506

 
alfa aesar™
1181506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.26
Chemical Name or Material Rhodium(III) chloride
Odor Odorless
Physical Form Powder
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula RhCl3
MDL Number MFCD00011204
CAS 10049-07-7
Merck Index 14,8188
Color Red-brown
Quantity 0.25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Rh 48.7% min.
RTECS Number VI9275000
TSCA TSCA
Name Note Anhydrous
Safety and Handling

Danger

GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.