missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min

$406.62 - $3,299.58

Chemical Identifiers

CAS 14284-92-5
Molecular Formula C15H24O6Rh
Molecular Weight (g/mol) 403.257
MDL Number MFCD00083144
InChI Key MBVAQOHBPXKYMF-MUCWUPSWSA-N
Synonym rhodium iii acetylacetonate-rh
PubChem CID 131675882
IUPAC Name (E)-4-hydroxypent-3-en-2-one;rhodium
SMILES CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Rh]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1056102
View Documents
Alfa Aesar™
1056102
0.5g
Each for $406.62
 
AA1056103
View Documents
Alfa Aesar™
1056103
1g
Each for $731.27
 
AA1056106
View Documents
Alfa Aesar™
1056106
5g
Each for $3,299.58
 
Description

Description

Specifications

Chemical Identifiers

14284-92-5
403.257
MBVAQOHBPXKYMF-MUCWUPSWSA-N
131675882
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Rh]
C15H24O6Rh
MFCD00083144
rhodium iii acetylacetonate-rh
(E)-4-hydroxypent-3-en-2-one;rhodium

Specifications

14284-92-5
MFCD00083144
Very soluble in chloroform; Soluble in alcohol; In soluble in water.
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Rh]
403.257
400.23
99.99%
Rh 25.2% min.
0.5g
(metals basis)
C15H24O6Rh
rhodium iii acetylacetonate-rh
MBVAQOHBPXKYMF-MUCWUPSWSA-N
(E)-4-hydroxypent-3-en-2-one;rhodium
131675882
Crystalline
Premion™
263°C to 264°C
Rhodium(III) 2,4-pentanedionate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 238-192-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS