missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium(II) Octanoate Dimer, TCI America™

Manufacturer:  Tci America R01611G

 View more versions of this product

Catalog No. R01611G



Specifications

Specifications

Rhodium(II) Octanoate Dimer
1g
C32H60O8Rh2
chloro 1,2,3-eta-3-phenyl-2-propenyl 1,3-bis 2,6-di-i-propylphenyl imidazol-2-ylidene palladium ii
CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].[Rh+2].[Rh+2]
778.635
778.64
Green
73482-96-9
MFCD00064724
FZXFNYFVNKTQSX-UHFFFAOYSA-J
octanoate;rhodium(2+)
131858392
Crystal-Powder at 20°C

Chemical Identifiers

73482-96-9
778.635
FZXFNYFVNKTQSX-UHFFFAOYSA-J
131858392
CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].[Rh+2].[Rh+2]
C32H60O8Rh2
MFCD00064724
chloro 1,2,3-eta-3-phenyl-2-propenyl 1,3-bis 2,6-di-i-propylphenyl imidazol-2-ylidene palladium ii
octanoate;rhodium(2+)
Safety and Handling

Safety and Handling

TSCA : No

Documents