Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(II) octanoate dimer

Chemical Identifiers

Chemical Name or Material Rhodium(II) octanoate dimer
CAS 73482-96-9
Linear Formula [Rh[CH3(CH2)6CO2]2]2
Formula Weight 778.63
MDL Number MFCD00064724
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3982503

  SDS
alfa aesar™
3982503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982506

  SDS
alfa aesar™
3982506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 778.63
Chemical Name or Material Rhodium(II) octanoate dimer
Physical Form Powder
Linear Formula [Rh[CH3(CH2)6CO2]2]2
MDL Number MFCD00064724
CAS 73482-96-9
Color Green
Quantity 1g
Solubility Information Soluble in hot alcohol, dichloromethane, toluene and acetic acid, slightly soluble in alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.