missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium(II) Acetate Dimer, TCI America™

Manufacturer:  Tci America R00691G

 View more versions of this product

Catalog No. R00691G



Specifications

Specifications

Rhodium(II) Acetate Dimer
240°C
15956-28-2
MFCD00003538
SYBXSZMNKDOUCA-UHFFFAOYSA-J
rhodium(2+);tetraacetate
122130469
Crystal-Powder at 20°C
Undesignated
1g
C8H12O8Rh2
acetic acid, rhodium 2+ salt
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Rh+2].[Rh+2]
441.987
441.99

Chemical Identifiers

15956-28-2
441.987
SYBXSZMNKDOUCA-UHFFFAOYSA-J
122130469
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Rh+2].[Rh+2]
C8H12O8Rh2
MFCD00003538
acetic acid, rhodium 2+ salt
rhodium(2+);tetraacetate
Safety and Handling

Safety and Handling

RTECSNumber : VI9361000

TSCA : No

Documents