Please login to your online account to display your discounted pricing

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  197612500

 View more versions of this product

Catalog No. AC197612500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 478.02
Physical Form Fine Powder
Quantity 250mg
Color Green to dark green
Assay 43% (RH)
Fieser 05,571; 08,434; 13,266; 15,278; 16,289; 17,298; 17,235
CAS 29998-99-0
Synonym Tetrakis[micron-(acetato-O:O')]diaquadirhodium
Health Hazard 1 Warning
Chemical Name or Material Rhodium(II) acetate dimer, dihydrate
Name Note ca 43% Rh
Molecular Formula C8H16O10Rh2
Linear Formula [(CH3CO2)2RhH2O]2
MDL Number MFCD00149212
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Rhodium(II) acetate dimer, dihydrate
Name Note ca 43% Rh
CAS 29998-99-0
Assay 43% (RH)
Molecular Formula C8H16O10Rh2
Linear Formula [(CH3CO2)2RhH2O]2
Formula Weight 478.02
MDL Number MFCD00149212
Synonym Tetrakis[micron-(acetato-O:O')]diaquadirhodium
Accessories

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents