missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 268632500

 View more versions of this product

Catalog No. AC268632500



Specifications

Specifications

Rhodium(II) acetate dimer
C8H12O8Rh2
acetic acid, rhodium 2+ salt, diacetoxyrhodium, dirhodium tetraacetate, rhodium acetate, brown, water soluble, rhodium diacetate, rhodium ii acetate, rhodium ii acetate dimer, rhodium ii acetate dimer dihydrate
SYBXSZMNKDOUCA-UHFFFAOYSA-J
rhodium(2+);tetraacetate
122130469
Micro-Crystalline Powder
Authentic
Solubility in water: insoluble. Other solubilities: moderately soluble in ethanol, acetone, acetonitri, le, dmso, thf
205.0°C
15956-28-2
15,8311
Solubility in water: insoluble. Other solubilities: moderately soluble in ethanol,acetone,acetonitri,le,dmso,thf
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Rh+2].[Rh+2]
441.98
441.98
ca 46% Rh
Glass bottle
Green
250mg

Chemical Identifiers

15956-28-2
441.98
acetic acid, rhodium 2+ salt, diacetoxyrhodium, dirhodium tetraacetate, rhodium acetate, brown, water soluble, rhodium diacetate, rhodium ii acetate, rhodium ii acetate dimer, rhodium ii acetate dimer dihydrate
rhodium(2+);tetraacetate
C8H12O8Rh2
SYBXSZMNKDOUCA-UHFFFAOYSA-J
122130469
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Rh+2].[Rh+2]
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 240-084-8

SDS Documents