missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium(II) acetate, dimer, 99.99% (metals basis), Rh 46.2% min., Alfa Aesar™ Premion™

For Use in Cyclopropanation of Alkenes

Manufacturer:  Alfa Aesar 1056001

 View more versions of this product

Catalog No. AA1056001



Specifications

Specifications

Rhodium(II) acetate, dimer
15956-28-2
C8H12O8Rh2
Green powder
SYBXSZMNKDOUCA-UHFFFAOYSA-J
rhodium(2+);tetraacetate
122130469
Powder
Green
Rh 46.2% min.
99.99% (metals basis)
MFCD00003538
rhodium ii acetate, rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium, rhodium ii acetate
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Rh+2].[Rh+2]
441.987
441.99
Odorless
0.25g

Chemical Identifiers

15956-28-2
441.987
SYBXSZMNKDOUCA-UHFFFAOYSA-J
122130469
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Rh+2].[Rh+2]
C8H12O8Rh2
MFCD00003538
rhodium ii acetate, rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium, rhodium ii acetate
rhodium(2+);tetraacetate
Documents