Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(II) acetate, dimer, 98+%

$76.19 - $885.08

Chemical Identifiers

Chemical Name or Material Rhodium(II) acetate
CAS 15956-28-2
Name Note dimer
Molecular Formula C8H12O8Rh2
Linear Formula [Rh(CO2CH3)2]2
Formula Weight 441.99
MDL Number MFCD00003538
Synonym Tetrakis(aceto)dirhodium(II); Dirhodium tetraacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL15152ME

  SDS
alfa aesar™
L15152ME
500mg Each for $250.91

AAL1515204

  SDS
alfa aesar™
L1515204
2g Each for $885.08

AAL15152MC

  SDS
alfa aesar™
L15152MC
100mg Each for $76.19
Description & Specifications

Specifications

Formula Weight 441.99
Assay ≥98%
Chemical Name or Material Rhodium(II) acetate
Odor Odorless
Name Note dimer
Molecular Formula C8H12O8Rh2
Linear Formula [Rh(CO2CH3)2]2
MDL Number MFCD00003538
CAS 15956-28-2
Synonym Tetrakis(aceto)dirhodium(II); Dirhodium tetraacetate
Melting Point 205°C
Quantity 500mg
RTECS Number VI9361000

Applications
Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ɑ-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

Solubility
Soluble in aqueous solution like water. Slightly soluble in ethanol

Notes
Stable under normal conditions.