Please login to your online account to display your discounted pricing

Rhodium on alumina, 5% Rh, powder, ACROS Organics™

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Formula Weight 102.91
MDL Number MFCD00011201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC199570250

  SDS
fisher scientific acros
199570250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC199570050

  SDS
fisher scientific acros
199570050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC199571000

  SDS
thermo fisher scientific
AC199571000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
CAS 7440-16-6
Molecular Formula Rh
Formula Weight 102.91
MDL Number MFCD00011201