missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium on activated carbon, 5% Rh, unreduced, dry, ACROS Organics™

Manufacturer:  Acros Organics 195440250

 View more versions of this product

Catalog No. AC195440250



Specifications

Specifications

Rhodium on activated carbon
5% Rh
Rh
01,982; 03,326; 04,418; 06,503; 08,433
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
[Rh]
102.906
23948
102.91
Typical particle size distribution: D50: 20 Micron, <8 Micron: 17%, <16 Micron: 39%,< 32 Micron: 67%, <64 Micron: 90%, <128 Micron: 99%
Black to Black-Gray
25g
7440-16-6,7440-44-0
MFCD00011201
15, 8310
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
Glass bottle
CHEBI:33359
Powder
5% (Rh)

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents