Learn More
Rhodium on activated carbon, 5% Rh, unreduced, dry, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 195440010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Black to Black-Gray | |
5% Rh | |
Rh | |
1 g | |
15, 8310 | |
MHOVAHRLVXNVSD-UHFFFAOYSA-N | |
rhodium | |
23948 | |
102.91 | |
Glass bottle | |
Powder |
Rhodium on activated carbon | |
7440-16-6,7440-44-0 | |
MFCD00011201 MFCD03458392 | |
01,982; 03,326; 04,418; 06,503; 08,433 | |
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac | |
[Rh] | |
102.91 | |
CHEBI:33359 | |
5% (Rh) | |
Typical particle size distribution: D50: 20 Micron, <8 Micron: 17%, <16 Micron: 39%,< 32 Micron: 67%, <64 Micron: 90%, <128 Micron: 99% |
Chemical Identifiers
7440-16-6 | |
102.91 | |
MHOVAHRLVXNVSD-UHFFFAOYSA-N | |
23948 | |
rhodium |
Rh | |
MFCD00011201 MFCD03458392 | |
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac | |
CHEBI:33359 | |
[Rh] |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only