missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rhodium acetate, ca. 40% Rh, brown, water soluble, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC270200050
Specifications
Rhodium acetate | |
42204-14-8 | |
Rh(CO2CH3)x | |
rhodium 3+ ion acetate | |
DVZJTSSQUUVXOC-UHFFFAOYSA-N | |
acetic acid;rhodium | |
3660445 | |
Powder | |
100ppm max. | |
100ppm max. | |
500ppm max. | |
100ppm max. | |
Glass bottle | |
250ppm max. | |
500ppm max. | |
100ppm max. | |
5g |
ca. 40% Rh | |
C2H3O2xRh | |
MFCD16556185 | |
Solubility in water: soluble | |
CC(=O)O.CC(=O)O.[Rh] | |
223.01 | |
130.96 | |
ca. 40% Rh | |
5000ppm max. | |
Authentic | |
250ppm max. | |
100ppm max. | |
250ppm max. | |
100ppm max. | |
250ppm max. | |
Brown |
Chemical Identifiers
42204-14-8 | |
223.01 | |
DVZJTSSQUUVXOC-UHFFFAOYSA-N | |
3660445 | |
CC(=O)O.CC(=O)O.[Rh] |
C2H3O2xRh | |
MFCD16556185 | |
rhodium 3+ ion acetate | |
acetic acid;rhodium |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Suspected of causing genetic defects.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical
GHS Signal Word: Warning
EINECSNumber : 255-707-9