Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 5% on alumina powder, Type 526, reduced

Chemical Identifiers

Chemical Name or Material Rhodium
CAS 7440-16-6
Name Note 5% on alumina powder, Type 526
MDL Number MFCD00011201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4712009

  SDS
alfa aesar™
4712009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4712018

  SDS
alfa aesar™
4712018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Chemical Name or Material Rhodium
CAS 7440-16-6
Name Note 5% on alumina powder, Type 526
TSCA TSCA
MDL Number MFCD00011201
Quantity 10g