missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium, 5% on alumina powder, reduced, Alfa Aesar™

$178.93 - $791.27

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.906
MDL Number MFCD00011201
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1177004
View Documents
Thermo Fisher Scientific
1177004
2g Each for $178.93
 
AA1177009
View Documents
Thermo Fisher Scientific
1177009
10g Each for $791.27
 
Specifications

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
CHEBI:33359
[Rh]

Specifications

2g
Black powder
MFCD00011201
Insoluble in water.
[Rh]
102.906
CHEBI:33359
Black
7440-16-6
Rh
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Powder
Rhodium
Safety and Handling

Safety and Handling

EINECSNumber : 231-125-0

TSCA : Yes

Recommended Storage : Ambient temperatures