missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodium, 5% on alumina powder, reduced, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1177004

 View more versions of this product

Catalog No. AA1177004



Specifications

Specifications

Rhodium
7440-16-6
Rh
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Powder
2g
Black powder
MFCD00011201
Insoluble in water.
[Rh]
102.906
CHEBI:33359
Black

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

EINECSNumber : 231-125-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents