Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 5% on activated carbon powder, Type C101038-5, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Rhodium
CAS 7440-16-6
Name Note 5% on activated carbon powder, Type C101038-5, Reduced, nominally 50% water wet
MDL Number MFCD00011201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1176103

 
alfa aesar™
1176103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1176109

 
alfa aesar™
1176109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1176118

 
alfa aesar™
1176118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Rhodium
TSCA TSCA
Name Note 5% on activated carbon powder, Type C101038-5, Reduced, nominally 50% water wet
Physical Form Powder
MDL Number MFCD00011201
CAS 7440-16-6
Color Black
Quantity 1g
Odor Odorless