missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 5% on activated carbon powder, Type C101038-5, reduced, nominally 50% water wet

Manufacturer:  Alfa Aesar™ 1176118

 View more versions of this product

Catalog No. AA1176118



Specifications

Specifications

Rhodium
5% on activated carbon powder, Type C101038-5, Reduced, nominally 50% water wet
Rh
black
[Rh]
102.906
CHEBI:33359
Odorless
50g
7440-16-6
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Powder
Black

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents