missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 5% on activated carbon powder, Type C101023-5, standard, reduced, nominally 50% water wet

Manufacturer:  Alfa Aesar™ 4542506

 View more versions of this product

Catalog No. AA4542506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Rhodium
7440-16-6
MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Powder
5g
5% on activated carbon powder, Type C101023-5, Standard, Reduced, Nominally 50% water wet
Rh
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
[Rh]
102.906
CHEBI:33359
Black

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD03458392
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents