missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 5% on activated carbon paste, C101023-5

Manufacturer:  Alfa Aesar™ 4486309

 View more versions of this product

Catalog No. AA4486309



Specifications

Specifications

Rhodium
7440-16-6
MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Paste
10g
5% on activated carbon paste, C101023-5
Rh
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
[Rh]
102.906
CHEBI:33359
Black

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD03458392
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents