missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 1% on alumina powder, reduced

$286.84 - $1,279.96

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.906
MDL Number MFCD00011201
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym black
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1176909 View Documents Alfa Aesar™
1176909
10g Each for $286.84
 
AA1176918 View Documents Alfa Aesar™
1176918
50g Each for $1,279.96
 
Specifications

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black
CHEBI:33359
[Rh]

Specifications

7440-16-6
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
10g
Rh
black
[Rh]
102.906
CHEBI:33359
Rhodium, 1% on alumina powder, Reduced
Safety and Handling

Safety and Handling

GHS H Statement
H335
May cause respiratory irritation.

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA