missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium, 1% on alumina powder, reduced

Manufacturer:  Alfa Aesar™ 1176918

 View more versions of this product

Catalog No. AA1176918



Specifications

Specifications

Rhodium
7440-16-6
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
50g
1% on alumina powder, Reduced
Rh
black
[Rh]
102.906
CHEBI:33359

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

GHS H Statement
H335
May cause respiratory irritation.

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents