missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodanine 99.0+%, TCI America™
$190.13 - $190.13
Chemical Identifiers
| CAS | 141-84-4 |
|---|---|
| Molecular Formula | C3H3NOS2 |
| Molecular Weight (g/mol) | 133.183 |
| MDL Number | MFCD00005488 |
| InChI Key | KIWUVOGUEXMXSV-UHFFFAOYSA-N |
| Synonym | rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline |
| PubChem CID | 1201546 |
| ChEBI | CHEBI:8830 |
| IUPAC Name | 2-sulfanylidene-1,3-thiazolidin-4-one |
| SMILES | C1C(=O)NC(=S)S1 |
Chemical Identifiers
| 141-84-4 | |
| 133.183 | |
| KIWUVOGUEXMXSV-UHFFFAOYSA-N | |
| 1201546 | |
| 2-sulfanylidene-1,3-thiazolidin-4-one |
| C3H3NOS2 | |
| MFCD00005488 | |
| rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline | |
| CHEBI:8830 | |
| C1C(=O)NC(=S)S1 |
Specifications
| 141-84-4 | |
| Yellow | |
| MFCD00005488 | |
| rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline | |
| C1C(=O)NC(=S)S1 | |
| 133.183 | |
| CHEBI:8830 | |
| ≥99.0% (T) | |
| Rhodanine |
| 168°C | |
| C3H3NOS2 | |
| 25 g | |
| KIWUVOGUEXMXSV-UHFFFAOYSA-N | |
| 2-sulfanylidene-1,3-thiazolidin-4-one | |
| 1201546 | |
| 133.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3410
RTECSNumber : VI7700000
TSCA : Yes