Please login to your online account to display your discounted pricing

Rhodanine, Reagent Grade, LabChem™

$94.83 - $94.83

Chemical Identifiers

Chemical Name or Material Rhodanine
CAS 141-84-4
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C3H3NOS2
Formula Weight 133.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC222408

 
lab chem
LC222408
Amber Glass 25g Each for $94.83
Description & Specifications

Specifications

Formula Weight 133.19
Packaging Amber Glass
Storage Information Room Temperature
Purity Grade Reagent Grade
Physical Form Powder
CAS 141-84-4
Solubility Information Soluble in water
Quantity 25g
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Specific Gravity 0.868g/cm3
Chemical Name or Material Rhodanine
EINECS Number 205-505-1
Density 0.868g/cm3
Linear Formula C3H3NOS2
Color Light yellow
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
f swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
Safety and Handling

Danger

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
f swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.