Please login to your online account to display your discounted pricing

Rhodanine Indicator, 0.02% (w/v), LabChem™

$51.49 - $51.49

Chemical Identifiers

Chemical Name or Material Rhodanine Indicator
CAS 7732-18-5
CAS Max % 99.98
CAS 141-84-4
CAS Max % 0.02
Linear Formula C3H3NOS2
Formula Weight 133.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC222507

 
lab chem
LC222507
Poly Bottle 125mL Each for $51.49
Description & Specifications

Specifications

Formula Weight 133.19
Chemical Name or Material Rhodanine Indicator
Quantity 125mL
Physical Form Liquid
Assay 0.02% (w/v)
Linear Formula C3H3NOS2
CAS 7732-18-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.98
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 141-84-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 1g/mL
CAS Max % 0.02
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1g/mL
Color Clear, slightly yellow
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature