missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodanine 99%, ACROS Organics™

Manufacturer:  Acros Organics 152601000

 View more versions of this product

Catalog No. AC152601000



Specifications

Specifications

Rhodanine
141-84-4
C3H3NOS2
27, 242
2-Thioxo-4-thiazolidinone
Authentic
2-sulfanylidene-1,3-thiazolidin-4-one
1201546
133.2
≥98.5%
167°C to 170°C
100g
99%
99%
MFCD00005488
15, 8308
KIWUVOGUEXMXSV-UHFFFAOYSA-N
C1C(=O)NC(=S)S1
133.183
CHEBI:8830
Fine Crystalline Powder
Glass bottle
Yellow to Beige

Chemical Identifiers

141-84-4
133.183
KIWUVOGUEXMXSV-UHFFFAOYSA-N
1201546
2-sulfanylidene-1,3-thiazolidin-4-one
C3H3NOS2
MFCD00005488
2-Thioxo-4-thiazolidinone
CHEBI:8830
C1C(=O)NC(=S)S1
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-505-1

RTECSNumber : VI7700000

TSCA : TSCA

SDS Documents