Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodanine, 98+%

$51.23 - $593.66

Chemical Identifiers

Chemical Name or Material Rhodanine
CAS 141-84-4
Molecular Formula C3H3NOS2
Formula Weight 133.19
MDL Number MFCD00005488
Beilstein 110701
Merck Index 14,8184
Synonym 4-Thiazolidinone-2-thione; 2-Thioxo-4-thiazolidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1829314

  SDS
alfa aesar™
A1829314
25g Each for $51.23

AAA1829322

  SDS
alfa aesar™
A1829322
100g Each for $146.61

AAA1829336

  SDS
alfa aesar™
A1829336
500g Each for $593.66
Description & Specifications

Specifications

Formula Weight 133.19
Chemical Name or Material Rhodanine
TSCA TSCA
Assay ≥98%
Molecular Formula C3H3NOS2
MDL Number MFCD00005488
CAS 141-84-4
Beilstein 110701
Merck Index 14,8184
Synonym 4-Thiazolidinone-2-thione; 2-Thioxo-4-thiazolidinone
Melting Point 166° to 171°C
Health Hazard 1 Danger
Quantity 25g
Odor Odorless
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280d-P305+P351+P338-P337+P313
Wear protective clothing and eye protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
RTECS Number VI7700000

Applications
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

Solubility
Soluble in water (2.25 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280d-P305+P351+P338-P337+P313
Wear protective clothing and eye protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.