missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodanine-3-propionic Acid 98.0+%, TCI America™

Manufacturer:  Tci America R006025G

 View more versions of this product

Catalog No. R006025G



Specifications

Specifications

Rhodanine-3-propionic Acid
25g
C6H7NO3S2
N-(2-Carboxyethyl)rhodanine
C1C(=O)N(C(=S)S1)CCC(=O)O
205.246
205.25
≥98.0% (T)
Undesignated
7025-19-6
MFCD00022558
SWAHCTPCIUXXTQ-UHFFFAOYSA-N
3-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)propanoic acid
81492
Crystal-Powder at 20°C

Chemical Identifiers

7025-19-6
205.246
SWAHCTPCIUXXTQ-UHFFFAOYSA-N
81492
C1C(=O)N(C(=S)S1)CCC(=O)O
C6H7NO3S2
MFCD00022558
N-(2-Carboxyethyl)rhodanine
3-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)propanoic acid
Safety and Handling

Safety and Handling

RTECSNumber : XJ6094060

TSCA : No

Documents