missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodanine-3-acetic Acid 98.0+%, TCI America™

Manufacturer:  Tci America R00545G

 View more versions of this product

Catalog No. R00545G



Specifications

Specifications

Rhodanine-3-acetic Acid
147°C
5718-83-2
MFCD00005491
JGRMXPSUZIYDRR-UHFFFAOYSA-N
2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
79793
Crystal-Powder at 20°C
Undesignated
5g
C5H5NO3S2
2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 3-carboxymethyl rhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acid, n-carboxymethyl rhodanine, n-carboxymethylrhodanine, rhodanine-3-acetic acid, rhodanine-n-acetic acid
C1C(=O)N(C(=S)S1)CC(=O)O
191.219
191.22
≥98.0% (T)

Chemical Identifiers

5718-83-2
191.219
JGRMXPSUZIYDRR-UHFFFAOYSA-N
79793
C1C(=O)N(C(=S)S1)CC(=O)O
C5H5NO3S2
MFCD00005491
2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 3-carboxymethyl rhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acid, n-carboxymethyl rhodanine, n-carboxymethylrhodanine, rhodanine-3-acetic acid, rhodanine-n-acetic acid
2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0223

TSCA : Yes

Documents