missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodanile Blue

$45.93 - $528.05

Chemical Identifiers

CAS 14969-56-3
Molecular Formula C48H48ClN5O3
Formula Weight 778.4
MDL Number MFCD00011937
Chemical Name or Material Rhodanile Blue
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1362203 SDS Alfa Aesar™
A1362203
1g Each for $45.93
AAA1362206 SDS Alfa Aesar™
A1362206
5g Each for $134.19
AAA1362214 SDS Alfa Aesar™
A1362214
25g Each for $528.05
Specifications Description & Specifications

Chemical Identifiers

14969-56-3
778.4
Rhodanile Blue
C48H48ClN5O3
MFCD00011937

Specifications

14969-56-3
778.4
1g
C48H48ClN5O3
MFCD00011937
Rhodanile Blue

Safety and Handling

GHS H Statement
H351-H319
Suspected of causing cancer.
Causes serious eye irritation.

GHS P Statement
P280-P281-P305+P351+P338-P308+P313-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning