Please login to your online account to display your discounted pricing

Rhodamine 6G, 99%, pure, laser grade, ACROS Organics™


Quantity 1g

Manufacturer: fisher scientific acros  419020010

 View more versions of this product

Catalog No. AC419020010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Rhodamine 6G
CAS 989-38-8
Purity Grade Laser Grade
Assay Percent Range 99%
Molecular Formula C28H31ClN2O3
Formula Weight 479.02
MDL Number MFCD00012665
Beilstein 18, V,12, 283
Synonym Basic Red 1
Quantity 1g

Specifications

Chemical Name or Material Rhodamine 6G
CAS 989-38-8
Purity Grade Laser Grade
Assay Percent Range 99%
Molecular Formula C28H31ClN2O3
Formula Weight 479.02
MDL Number MFCD00012665
Synonym Basic Red 1