Please login to your online account to display your discounted pricing

Rhodamine 6G, 99%, pure, laser grade, ACROS Organics™

$101.09 - $101.09

Chemical Identifiers

Chemical Name or Material Rhodamine 6G
CAS 989-38-8
Purity Grade Laser Grade
Assay Percent Range 99%
Molecular Formula C28H31ClN2O3
Formula Weight 479.02
MDL Number MFCD00012665
Synonym Basic Red 1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC419025000

  SDS
fisher scientific acros
419025000
500mg Each for $101.09

AC419020010

  SDS
fisher scientific acros
419020010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC419020050

  SDS
thermo fisher scientific
AC419020050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Rhodamine 6G
CAS 989-38-8
Purity Grade Laser Grade
Assay Percent Range 99%
Molecular Formula C28H31ClN2O3
Formula Weight 479.02
MDL Number MFCD00012665
Beilstein 18, V,12, 283
Synonym Basic Red 1
Quantity 500mg