Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodamine 110 bis(Z-L-isoleucyl-L-prolyl-L-arginine amide) dihydrochloride

Chemical Identifiers

Chemical Name or Material Rhodamine 110 bis(Z-L-isoleucyl-L-prolyl-L-arginine amide) dihydrochloride
Formula Weight 1404.46
MDL Number MFCD00467932
Synonym (Z-Ile-Pro-Arg)2-R110; BZiPAR
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66754EXD

 
alfa aesar™
J66754EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66754MCR

 
alfa aesar™
J66754MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1404.46
Chemical Name or Material Rhodamine 110 bis(Z-L-isoleucyl-L-prolyl-L-arginine amide) dihydrochloride
MDL Number MFCD00467932
Physical Form Solid
Synonym (Z-Ile-Pro-Arg)2-R110; BZiPAR
Storage Note 1 -30° to -10°C
Quantity 0.5mg
Color White to off-white
Safety and Handling

-30° to -10°C