Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodamine 110 bis(succinoyl-L-alanyl-L-alanyl-L-prolyl-L-phenylalanyl amide)

Chemical Identifiers

Chemical Name or Material Rhodamine 110 bis(succinoyl-L-alanyl-L-alanyl-L-prolyl-L-phenylalanyl amide)
Formula Weight 1303.39
MDL Number MFCD03428407
Synonym (Suc-Ala-Ala-Pro-Phe)2-R110
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66124EXD

 
alfa aesar™
J66124EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66124MCR

 
alfa aesar™
J66124MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1303.39
Chemical Name or Material Rhodamine 110 bis(succinoyl-L-alanyl-L-alanyl-L-prolyl-L-phenylalanyl amide)
Quantity 0.5mg
Physical Form Solid
MDL Number MFCD03428407
Synonym (Suc-Ala-Ala-Pro-Phe)2-R110
Storage Note 1 -30° to -10°C
Color White to off-white
Safety and Handling

-30° to -10°C