Please login to your online account to display your discounted pricing

eBiosciencetrade; Rhesus Fc Receptor Binding Inhibitor, Purified

Can be used to inhibit the non-specific Fc receptor (FcR)-mediated binding of mouse monoclonal antibodies used for flow cytometric analysis of rhesus tissue.

$303.05 - $303.05

Specifications

Host Species Rhesus
Formulation Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C
Concentration 5μL (10μg)/test
For Use With (Application) Flow Cytometric Analysis
View More Specs

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122773

 
ebioscience
LS14916542
100 tests Each for $303.05
Description & Specifications

Specifications

Host Species Rhesus
Formulation Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C
Concentration 5μL (10μg)/test
For Use With (Application) Flow Cytometric Analysis