missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium wire, 1.0mm (0.04 in.) dia., 99.97% (metals basis)

Manufacturer:  Alfa Aesar™ 14169BS

 View more versions of this product

Catalog No. AA14169BS



Specifications

Specifications

Rhenium wire
10cm
7440-15-5
Re
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
[Re]
Odorless
23947
Wire
≈16.52g/m
1.0mm (0.04 in.) dia.
(metals basis)
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
186.207
CHEBI:49882
99.97%

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents