missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium Wire, 0.5mm (0.02 in.) dia., 99.97% (Metals basis), Alfa Aesar™

Form: Approx.4.13g/m

Manufacturer:  Alfa Aesar 10312BU

 View more versions of this product

Catalog No. AA10312BU



Specifications

Specifications

Rhenium
7440-15-5
21.04g/cm3
Re
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
[Re]
Odorless
23947
Wire, 0.5mm (0.02 in.) dia.
25cm
Silver gray wire
99.97% (Metals basis)
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
186.207
CHEBI:49882
Silver to Gray

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]
Documents