missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium(VII) oxide, 99.995% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4553804

 View more versions of this product

Catalog No. AA4553804



Specifications

Specifications

Rhenium(VII) oxide
O7Re2
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
O=[Re](=O)(=O)O[Re](=O)(=O)=O
484.407
484.4
Odorless
2g
1314-68-7
MFCD00011199
NBGOSNNAAHDRLK-UHFFFAOYSA-N
trioxo(trioxorheniooxy)rhenium
123106
Crystals
Yellow

Chemical Identifiers

1314-68-7
484.407
NBGOSNNAAHDRLK-UHFFFAOYSA-N
123106
O=[Re](=O)(=O)O[Re](=O)(=O)=O
O7Re2
MFCD00011199
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
trioxo(trioxorheniooxy)rhenium
Safety and Handling

Safety and Handling

H314-H318-H272: Causes severe skin burns and eye damage. Causes serious eye damage. May intensify fire; oxidizer.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 215-241-9

SDS Documents