Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium(VII) oxide, 99.9% (metals basis)

As a catalyst for oxidation/reduction reactions

Chemical Identifiers

Chemical Name or Material Rhenium(VII) oxide
CAS 1314-68-7
Linear Formula Re2O7
Formula Weight 484.4
MDL Number MFCD00011199
UN Number UN3085
Merck Index 14,8178
Synonym Rhenium heptoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4046804

 
alfa aesar™
4046804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4046809

 
alfa aesar™
4046809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4046818

 
alfa aesar™
4046818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 484.4
Packaging Ampule
Chemical Name or Material Rhenium(VII) oxide
TSCA TSCA
Odor Odorless
Physical Form Crystalline
Assay 99.9%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
Linear Formula Re2O7
MDL Number MFCD00011199
CAS 1314-68-7
UN Number UN3085
Merck Index 14,8178
Synonym Rhenium heptoxide
Boiling Point 360°C
Color Yellow to pale green
Melting Point 297°C
Quantity 2g
Solubility Information Dissolves in water to form HReO4; Very soluble in alcohol; Soluble in alkalies and acids
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H272
Causes severe skin burns and eye damage.
Causes serious eye damage.
May intensify fire
oxidizer.
Health Hazard 3 GHS P Statement
P221-P280-P305+P351+P338-P309-P310
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Storage Note 1
  • Ampouled under argon
Density 6.103g/mL
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P221-P280-P305+P351+P338-P309-P310
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H272
Causes severe skin burns and eye damage.
Causes serious eye damage.
May intensify fire
oxidizer.

  • Ampouled under argon