missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium Sponge/powder, 99.95% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1207304

 View more versions of this product

Catalog No. AA1207304



Specifications

Specifications

Rhenium
7440-16-6
99.95% (Metals basis)
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
102.906
CHEBI:33359
Gray
2g
gray sponge/powder
Rh
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
[Rh]
Odorless
23948
Sponge/Powder

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-125-0

Documents