missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium Sponge/powder, 99.95% (Metals basis), Alfa Aesar™

Chemical Identifiers

CAS 7440-16-6
Physical Form Sponge/Powder
Assay Percent Range 99.95% (Metals basis)
MDL Number MFCD00011201
Chemical Name or Material Rhenium
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1207304 SDS Alfa Aesar
1207304
2g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7440-16-6
99.95% (Metals basis)
Rhenium
Sponge/Powder
MFCD00011201

Specifications

2g
gray sponge/powder
99.95% (Metals basis)
Odorless
Rhenium
7440-16-6
Sponge/Powder
MFCD00011201
Gray