Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium silicide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Rhenium silicide
CAS 12038-66-3
Formula Weight 242.37
MDL Number MFCD00148218
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3638304

  SDS
alfa aesar™
3638304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.37
Chemical Name or Material Rhenium silicide
TSCA TSCA
Mesh Size −80 Mesh
Odor Odorless
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00148218
CAS 12038-66-3
Quantity 2g