missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium powder, 99.99%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1031304

 View more versions of this product

Catalog No. AA1031304



Specifications

Specifications

Rhenium
7440-15-5
99.99% (Metals basis)
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
Powder
2g
Powder
Re
7yhu292iny
[Re]
186.207
CHEBI:49882

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
7yhu292iny
CHEBI:49882
[Re]
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-124-5

Documents