missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium powder, -22 mesh, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1067303

 View more versions of this product

Catalog No. AA1067303



Specifications

Specifications

Rhenium powder
7440-15-5
Re
UN3089
7yhu292iny, atom, foil, hydride, ion re7, pellets, renio, rhenium, elementar, rhenium, ion re7, unii-7yhu292iny
[Re]
186.207
CHEBI:49882
99.999%
1g
(metals basis)
MFCD00011195
−22 Mesh
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
Powder
Puratronic™

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
7yhu292iny, atom, foil, hydride, ion re7, pellets, renio, rhenium, elementar, rhenium, ion re7, unii-7yhu292iny
CHEBI:49882
[Re]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-124-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents