Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium, plasma standard solution, Specpure™, Re 10,000μg/mL

$342.04 - $342.04

Chemical Identifiers

Chemical Name or Material Rhenium plasma standard solution
Name Note Re 10000μg/mL
Purity Grade Specpure™
MDL Number MFCD00011195
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14416AD

  SDS
alfa aesar™
14416AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14416AE

  SDS
alfa aesar™
14416AE
100mL Each for $342.04

AA14416AP

  SDS
alfa aesar™
14416AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note Re 10000μg/mL
Quantity 50mL
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Purity Grade Specpure™
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Physical Form Liquid
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
UN Number UN3264
Health Hazard 1 Danger
Chemical Name or Material Rhenium plasma standard solution
MDL Number MFCD00011195
Concentration Re in 5% HNO3
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.